Assay Details
Target Gene Details
Entrez Gene ID: | 10063 |
Gene Name: | COX17, cytochrome c oxidase copper chaperone |
Gene Aliases: |
- |
Location: |
Chr.3:119659975-119677441 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COX17 | NM_005694.1 | NP_005685.1 | ||
| AK054900.1 | ||||
| AK095366.1 | 2 | 695 | ||
| BC010933.1 | AAH10933.1 | |||
| BC105280.1 | AAI05281.1 | |||
| L77701.1 | AAA98114.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1014394 | Chr.3:119641659 - 119693428 on Build GRCh38 | Loss |
|
| esv3568977 | Chr.3:119657752 - 119694492 on Build GRCh38 | Loss |
|
| nsv460834 | Chr.3:119665316 - 119740004 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map