Assay Details
Target Gene Details
Entrez Gene ID: | 90102 |
Gene Name: | pleckstrin homology like domain family B member 2 |
Gene Aliases: |
LL5b, LL5beta |
Location: |
Chr.3:111732480-111976517 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHLDB2 | NM_001134437.1 | NP_001127909.1 | ||
| AK296841.1 | ||||
| AK307491.1 | ||||
| CR749654.1 | CAH18447.1 | |||
| DA447763.1 |
Target Gene Details
Entrez Gene ID: | 257068 |
Gene Name: | phosphatidylinositol specific phospholipase C X domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.3:111674676-111846447 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLCXD2 | NM_001185106.1 | NP_001172035.1 | ||
| NM_153268.3 | NP_695000.1 | |||
| AK056141.1 | BAB71103.1 | |||
| BC121155.1 | ||||
| BC121156.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv591275 | Chr.3:110918624 - 111782250 on Build GRCh38 | Gain |
|
| nsv591290 | Chr.3:111708608 - 111743723 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map