Assay Details
Target Gene Details
Entrez Gene ID: | 7324 |
Gene Name: | ubiquitin conjugating enzyme E2 E1 |
Gene Aliases: |
UBCH6 |
Location: |
Chr.3:23805893-23891640 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE2E1 | NM_001202476.1 | NP_001189405.1 | ||
| NM_003341.4 | NP_003332.1 | |||
| NM_182666.2 | NP_872607.1 | |||
| XM_005265431.4 | XP_005265488.1 | |||
| AK295354.1 | ||||
| AK314854.1 | ||||
| BC009139.1 | AAH09139.1 | |||
| BF698653.1 | ||||
| BI223271.1 | ||||
| DB478686.1 | ||||
| DB499676.1 | ||||
| X92963.1 | CAA63539.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834637 | Chr.3:23662437 - 23844653 on Build GRCh38 | Loss |
|
| nsv1005186 | Chr.3:21989661 - 24089392 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73822595] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map