Assay Details
Target Gene Details
Entrez Gene ID: | 101927854 |
Gene Name: | uncharacterized LOC101927854 |
Gene Aliases: |
- |
Location: |
Chr.3:24151324-24189956 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927854 | NR_121667.1 | |||
| CB994391.1 | ||||
| KJ999766.1 |
Target Gene Details
Entrez Gene ID: | 7068 |
Gene Name: | thyroid hormone receptor beta |
Gene Aliases: |
C-ERBA-2, C-ERBA-BETA, ERBA2, GRTH, NR1A2, PRTH, THR1, THRB1, THRB2 |
Location: |
Chr.3:24117153-24495281 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834639 | Chr.3:23995622 - 24192434 on Build GRCh38 | Gain |
|
| nsv3733 | Chr.3:24175086 - 24220258 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map