Assay Details
Target Gene Details
Entrez Gene ID: | 112616 |
Gene Name: | CKLF like MARVEL transmembrane domain containing 7 |
Gene Aliases: |
CKLFSF7 |
Location: |
Chr.3:32391671-32455528 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CMTM7 | NM_138410.3 | NP_612419.1 | ||
| NM_181472.2 | NP_852137.1 | |||
| XM_011533319.2 | XP_011531621.1 | |||
| XM_017005646.1 | XP_016861135.1 | |||
| AF479263.2 | AAN73041.1 | |||
| AK308878.1 | ||||
| AL832450.1 | 1 | 680 | CAD38626.1 | |
| AY174113.1 | AAO44955.1 | |||
| BC010116.2 | AAH10116.1 | |||
| DQ132881.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1003285 | Chr.3:31386933 - 33172886 on Build GRCh38 | Gain |
|
| nsv834652 | Chr.3:32375792 - 32564922 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map