Assay Details
Target Gene Details
Entrez Gene ID: | 344905 |
Gene Name: | ATPase 13A5 |
Gene Aliases: |
- |
Location: |
Chr.3:193274789-193378824 on Build GRCh38 |
Assay Gene Location: | Within Intron 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP13A5 | NM_198505.2 | NP_940907.2 | ||
| XM_017006304.1 | XP_016861793.1 | |||
| XM_017006305.1 | XP_016861794.1 | |||
| AY358667.1 | AAQ89030.1 | |||
| AY823163.1 | AAX24103.1 |
Target Gene Details
Entrez Gene ID: | 57110 |
Gene Name: | HRAS like suppressor |
Gene Aliases: |
A-C1, H-REV107, HRASLS1, HRSL1, HSD28 |
Location: |
Chr.3:193241128-193281639 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HRASLS | XM_011513035.2 | XP_011511337.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1003505 | Chr.3:193168634 - 193574329 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map