Assay Details
Target Gene Details
Entrez Gene ID: | 8850 |
Gene Name: | lysine acetyltransferase 2B |
Gene Aliases: |
CAF, P/CAF, PCAF |
Location: |
Chr.3:20040032-20154404 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KAT2B | NM_003884.4 | NP_003875.3 | ||
| XM_005265528.4 | XP_005265585.1 | |||
| BC060823.1 | AAH60823.1 | |||
| BC070075.1 | AAH70075.1 | |||
| U57317.2 | AAC50890.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893667 | Chr.3:19470582 - 20214665 on Build GRCh38 | Gain |
|
| esv3893669 | Chr.3:20001342 - 20074585 on Build GRCh38 | Loss |
|
| esv35078 | Chr.3:19513779 - 20728244 on Build GRCh38 | Gain |
|
| nsv997486 | Chr.3:19965210 - 20072497 on Build GRCh38 | Gain |
|
| esv23068 | Chr.3:19494161 - 20597009 on Build GRCh38 | Gain+Loss |
|
| nsv834627 | Chr.3:20026623 - 20186483 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map