Assay Details
Target Gene Details
Entrez Gene ID: | 523 |
Gene Name: | ATPase H+ transporting V1 subunit A |
Gene Aliases: |
ATP6A1, ATP6V1A1, HO68, VA68, VPP2, Vma1 |
Location: |
Chr.3:113747019-113812058 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP6V1A | NM_001690.3 | NP_001681.2 | ||
| AF113129.1 | AAF14870.1 | |||
| AK293804.1 | ||||
| AK295150.1 | ||||
| AK314779.1 | ||||
| AL514316.3 | ||||
| BC013138.1 | AAH13138.1 | |||
| DC350361.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2588841 | Chr.3:113761339 - 113762890 on Build GRCh38 | Deletion |
|
| nsv1013506 | Chr.3:113171761 - 113962104 on Build GRCh38 | Gain |
|
| nsv829678 | Chr.3:113727330 - 113914206 on Build GRCh38 | Loss |
|
| nsv1013746 | Chr.3:113741315 - 113927810 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map