Assay Details
Target Gene Details
Entrez Gene ID: | 131377 |
Gene Name: | kelch like family member 40 |
Gene Aliases: |
KBTBD5, NEM8, SRYP, SYRP |
Location: |
Chr.3:42685519-42692446 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLHL40 | NM_152393.3 | NP_689606.2 | ||
| AK056577.1 | BAB71222.1 | |||
| AK291294.1 | ||||
| AK300682.1 | ||||
| AY176040.1 | AAO22141.1 | |||
| AY177390.1 | AAO06908.1 | |||
| BC110491.2 | AAI10492.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv954472 | Chr.3:42683809 - 42713908 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map