Assay Details
Target Gene Details
Entrez Gene ID: | 100506826 |
Gene Name: | MYLK antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:123585513-123630821 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYLK-AS1 | NR_038266.2 | |||
| NR_121654.1 | ||||
| DA531919.1 | ||||
| DA714112.1 | ||||
| DB499540.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv591428 | Chr.3:123608828 - 124139902 on Build GRCh38 | Gain |
|
| nsv829708 | Chr.3:123519589 - 123690957 on Build GRCh38 | Gain |
|
| nsv829709 | Chr.3:123554281 - 123748120 on Build GRCh38 | Loss |
|

Genomic Map