Assay Details
Target Gene Details
Entrez Gene ID: | 55171 |
Gene Name: | TBCC domain containing 1 |
Gene Aliases: |
- |
Location: |
Chr.3:186546067-186570543 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBCCD1 | NM_001134415.1 | NP_001127887.1 | ||
| NM_001286749.1 | NP_001273678.1 | |||
| AK124362.1 | ||||
| DB033833.1 |
More Information
Set Membership: |
|

Genomic Map