Assay Details
Target Gene Details
Entrez Gene ID: | 54681 |
Gene Name: | prolyl 4-hydroxylase, transmembrane |
Gene Aliases: |
EGLN4, HIFPH4, P4H-TM, PH-4, PH4, PHD4 |
Location: |
Chr.3:48989908-49007149 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
P4HTM | NM_177938.2 | NP_808807.2 | ||
NM_177939.2 | NP_808808.1 | |||
AK000269.1 | BAA91045.1 | |||
AL713728.1 | CAD28518.2 | |||
AY198406.1 | AAO43431.1 | |||
BC011710.2 | AAH11710.3 | |||
BC060321.1 | AAH60321.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
