Assay Details
Target Gene Details
Entrez Gene ID: | 344805 |
Gene Name: | transmembrane protease, serine 7 |
Gene Aliases: |
- |
Location: |
Chr.3:112039067-112081269 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMPRSS7 | XM_011512754.1 | XP_011511056.1 |
More Information
Set Membership: |
|

Genomic Map