Assay Details
Target Gene Details
Entrez Gene ID: | 9223 |
Gene Name: | membrane associated guanylate kinase, WW and PDZ domain containing 1 |
Gene Aliases: |
AIP-3, AIP3, BAIAP1, BAP-1, BAP1, MAGI-1, Magi1d, TNRC19, WWP3 |
Location: |
Chr.3:65353525-66038836 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Gene Details
Entrez Gene ID: | 100873983 |
Gene Name: | MAGI1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:65893816-65925297 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAGI1-AS1 | NR_046575.1 | |||
| BX089101.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv460697 | Chr.3:65861250 - 65938065 on Build GRCh38 | Loss |
|
| nsv590546 | Chr.3:65487766 - 67232113 on Build GRCh38 | Gain |
|
| nsv590548 | Chr.3:65811865 - 65933794 on Build GRCh38 | Loss |
|
| dgv4769n100 | Chr.3:65724045 - 65902469 on Build GRCh38 | Gain |
|
| nsv1000854 | Chr.3:65842708 - 65916742 on Build GRCh38 | Loss |
|
| nsv1012099 | Chr.3:65899988 - 65979750 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map