Assay Details
Target Gene Details
Entrez Gene ID: | 131890 |
Gene Name: | G protein-coupled receptor kinase 7 |
Gene Aliases: |
GPRK7 |
Location: |
Chr.3:141763127-141817373 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRK7 | NM_139209.2 | NP_631948.1 | ||
| XM_017005717.1 | XP_016861206.1 | |||
| AF282269.1 | AAL33880.1 | |||
| AF439409.1 | AAL48216.1 |
More Information
Set Membership: |
|

Genomic Map