Assay Details
Target Gene Details
Entrez Gene ID: | 8626 |
Gene Name: | tumor protein p63 |
Gene Aliases: |
AIS, B(p51A), B(p51B), EEC3, KET, LMS, NBP, OFC8, RHS, SHFM4, TP53CP, TP53L, TP73L, p40, p51, p53CP, p63, p73H, p73L |
Location: |
Chr.3:189597115-189897279 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TP63 | NM_001114978.1 | NP_001108450.1 | ||
| NM_001114979.1 | NP_001108451.1 | |||
| NM_003722.4 | NP_003713.3 | |||
| XM_005247843.3 | XP_005247900.1 | |||
| XM_005247844.4 | XP_005247901.1 | |||
| XM_011513251.2 | XP_011511553.1 | |||
| XM_011513252.2 | XP_011511554.1 | |||
| AB016072.1 | BAA32592.1 | |||
| AB016073.1 | BAA32593.1 | |||
| AF116771.1 | AAF61624.1 | |||
| BC039815.1 | AAH39815.1 | |||
| KU178435.1 | ||||
| KU178436.1 | ||||
| Y16961.1 | CAA76562.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508265 | Chr.3:189636239 - 189660195 on Build GRCh38 | Deletion |
|
| nsv4167 | Chr.3:189638804 - 189662392 on Build GRCh38 | Deletion |
|
| nsv592764 | Chr.3:189639152 - 189650568 on Build GRCh38 | Loss |
|
| nsv1014449 | Chr.3:189636314 - 189753850 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map