Assay Details
Target Gene Details
Entrez Gene ID: | 80218 |
Gene Name: | N(alpha)-acetyltransferase 50, NatE catalytic subunit |
Gene Aliases: |
MAK3, NAT13, NAT13P, NAT5, NAT5P, SAN |
Location: |
Chr.3:113716460-113746299 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAA50 | NM_001308445.1 | 5 | 5983 | NP_001295374.1 |
| NM_025146.3 | 5 | 5986 | NP_079422.1 | |
| BX102492.1 | 1 | 188 | ||
| H16725.1 | 1 | 189 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1013506 | Chr.3:113171761 - 113962104 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72952659] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map