Assay Details
Target Gene Details
Entrez Gene ID: | 93973 |
Gene Name: | ARP8 actin-related protein 8 homolog |
Gene Aliases: |
ARP8, INO80N, hArp8 |
Location: |
Chr.3:53863096-53882202 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACTR8 | XM_005265587.4 | 14 | 3319 | XP_005265644.1 |
Target Gene Details
Entrez Gene ID: | 55540 |
Gene Name: | interleukin 17 receptor B |
Gene Aliases: |
CRL4, EVI27, IL17BR, IL17RH1 |
Location: |
Chr.3:53846494-53866526 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL17RB | NM_018725.3 | NP_061195.2 | ||
| XM_005265310.4 | XP_005265367.1 | |||
| XM_005265311.4 | XP_005265368.1 | |||
| XM_005265312.4 | XP_005265369.1 | |||
| XM_011533941.2 | XP_011532243.1 | |||
| XM_017006804.1 | XP_016862293.1 | |||
| XM_017006805.1 | XP_016862294.1 | |||
| XM_017006806.1 | XP_016862295.1 | |||
| XM_017006807.1 | XP_016862296.1 | |||
| AF208110.1 | AAF86051.1 | |||
| AF208111.1 | AAF86052.1 | |||
| AF212365.1 | AAF78776.1 | |||
| AF250309.1 | AAK37428.1 | |||
| AK307749.1 | ||||
| AY359122.1 | AAQ89956.1 | |||
| BC000980.1 | AAH00980.1 | |||
| BI458542.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1376e212 | Chr.3:53859682 - 53883597 on Build GRCh38 | Loss |
|
| nsv1013783 | Chr.3:53660391 - 53900754 on Build GRCh38 | Gain |
|
| esv3575570 | Chr.3:53721382 - 53883305 on Build GRCh38 | Gain |
|
| nsv1006990 | Chr.3:53815483 - 54251185 on Build GRCh38 | Gain |
|
| nsv1003621 | Chr.3:53371619 - 54391295 on Build GRCh38 | Loss |
|
| dgv4747n100 | Chr.3:53812714 - 53900754 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map