Assay Details
Target Gene Details
Entrez Gene ID: | 27032 |
Gene Name: | ATPase secretory pathway Ca2+ transporting 1 |
Gene Aliases: |
ATP2C1A, BCPM, HHD, PMR1, SPCA1, hSPCA1 |
Location: |
Chr.3:130850500-131016712 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP2C1 | NM_001199180.1 | 1 | 211 | NP_001186109.1 |
| NM_001199181.1 | 1 | 211 | NP_001186110.1 | |
| NM_001199182.1 | 1 | 211 | NP_001186111.1 | |
| XM_005247354.2 | 1 | 236 | XP_005247411.1 | |
| AK296470.1 | 1 | 138 | ||
| AK299945.1 | ||||
| AK304374.1 | ||||
| DC422788.1 | 1 | 211 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1005160 | Chr.3:130723453 - 131010147 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map