Assay Details
Target Gene Details
Entrez Gene ID: | 91653 |
Gene Name: | BOC cell adhesion associated, oncogene regulated |
Gene Aliases: |
CDON2 |
Location: |
Chr.3:113211003-113287463 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BOC | XM_017007446.1 | XP_016862935.1 | ||
| XM_017007448.1 | XP_016862937.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1013506 | Chr.3:113171761 - 113962104 on Build GRCh38 | Gain |
|
| nsv1144997 | Chr.3:113209953 - 113212253 on Build GRCh38 | Deletion |
|
| nsv829677 | Chr.3:113138228 - 113320140 on Build GRCh38 | Gain |
|
| nsv591293 | Chr.3:112266260 - 113293950 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map