Assay Details
Target Gene Details
Entrez Gene ID: | 10869 |
Gene Name: | ubiquitin specific peptidase 19 |
Gene Aliases: |
ZMYND9 |
Location: |
Chr.3:49108046-49120938 on Build GRCh38 |
Assay Gene Location: | Within Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP19 | NM_001199161.1 | 27 | 4625 | NP_001186090.1 |
| NM_001199162.1 | 27 | 4589 | NP_001186091.1 | |
| XM_005264829.2 | 27 | 4626 | XP_005264886.1 | |
| XM_005264831.2 | 26 | 4272 | XP_005264888.1 | |
| XM_006712948.2 | 27 | 4614 | XP_006713011.1 | |
| XM_006712950.2 | 27 | 4575 | XP_006713013.1 | |
| XM_006712951.2 | 27 | 4572 | XP_006713014.1 | |
| XM_006712952.2 | 27 | 4569 | XP_006713015.1 | |
| XM_017005618.1 | 27 | 4620 | XP_016861107.1 | |
| XM_017005619.1 | 27 | 4617 | XP_016861108.1 | |
| XM_017005620.1 | 27 | 4611 | XP_016861109.1 | |
| XM_017005622.1 | 27 | 4581 | XP_016861111.1 | |
| XM_017005624.1 | 27 | 4464 | XP_016861113.1 | |
| XM_017005630.1 | 26 | 4323 | XP_016861119.1 | |
| XM_017005631.1 | 26 | 4317 | XP_016861120.1 | |
| XM_017005632.1 | 26 | 4317 | XP_016861121.1 | |
| XM_017005633.1 | 26 | 4311 | XP_016861122.1 | |
| XM_017005634.1 | 26 | 4278 | XP_016861123.1 | |
| AK300425.1 | 27 | 4443 | ||
| BC048269.1 | 8 | 1464 | AAH48269.1 | |
| BC082241.1 | 24 | 3692 | AAH82241.1 | |
| BC106029.1 | 15 | 2553 | AAI06030.1 | |
| BC142660.1 | 27 | 4495 | ||
| BC142727.1 | 27 | 4501 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834686 | Chr.3:49068614 - 49233123 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map