Assay Details
Target Gene Details
| Entrez Gene ID: | 10201 | 
| Gene Name: | NME/NM23 nucleoside diphosphate kinase 6 | 
| Gene Aliases: | IPIA-ALPHA, NDK 6, NM23-H6 | 
| Location: | Chr.3:48289369-48302259 on Build GRCh38 | 
| Assay Gene Location: | Within Exon 2 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv834683 | Chr.3:48260014 - 48451855 on Build GRCh38 | Gain |  ATRIP  TMA7  ZNF589  PLXNB1  NME6  SPINK8  FCF1P2  CCDC51  MIR2115  FBXW12 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top