Assay Details
Target Gene Details
Entrez Gene ID: | 3708 |
Gene Name: | inositol 1,4,5-trisphosphate receptor type 1 |
Gene Aliases: |
ACV, CLA4, INSP3R1, IP3R, IP3R1, PPP1R94, SCA15, SCA16, SCA29 |
Location: |
Chr.3:4493348-4847840 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422450 | Chr.3:4467187 - 4614541 on Build GRCh38 | Deletion |
|
| nsv834605 | Chr.3:4508590 - 4643041 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map