Assay Details
Target Gene Details
Entrez Gene ID: | 253559 |
Gene Name: | cell adhesion molecule 2 |
Gene Aliases: |
IGSF4D, NECL3, Necl-3, SynCAM 2, synCAM2 |
Location: |
Chr.3:84958982-86074429 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CADM2 | NM_001167674.1 | NP_001161146.1 | ||
| NM_001167675.1 | NP_001161147.1 | |||
| XM_006713081.3 | XP_006713144.1 | |||
| XM_006713082.3 | XP_006713145.1 | |||
| XM_017006062.1 | XP_016861551.1 | |||
| AB646739.1 | ||||
| AK295885.1 | ||||
| AL834270.1 | CAD38945.1 | |||
| BC105999.1 | AAI06000.1 | |||
| BC112136.1 | AAI12137.1 | |||
| BC112138.1 | AAI12139.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1002058 | Chr.3:84707893 - 85340726 on Build GRCh38 | Gain |
|
| nsv1010140 | Chr.3:84947861 - 84985130 on Build GRCh38 | Gain |
|
| nsv1000793 | Chr.3:84971301 - 85002688 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map