Assay Details
Target Gene Details
Entrez Gene ID: | 79750 |
Gene Name: | zinc finger protein 385D |
Gene Aliases: |
ZNF659 |
Location: |
Chr.3:21418419-22372641 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF385D | XM_011534122.1 | XP_011532424.1 | ||
| XM_011534123.2 | XP_011532425.1 | |||
| XM_011534124.2 | XP_011532426.1 | |||
| XM_017007191.1 | XP_016862680.1 | |||
| XM_017007192.1 | XP_016862681.1 | |||
| XM_017007194.1 | XP_016862683.1 | |||
| XM_017007199.1 | XP_016862688.1 | |||
| XM_017007203.1 | XP_016862692.1 | |||
| AK310298.1 |
Target Gene Details
Entrez Gene ID: | 100874221 |
Gene Name: | ZNF385D antisense RNA 2 |
Gene Aliases: |
- |
Location: |
Chr.3:21957139-21979828 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF385D-AS2 | NR_046876.1 | |||
| AI200816.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834633 | Chr.3:21876772 - 22046846 on Build GRCh38 | Loss |
|
| dgv4696n100 | Chr.3:21942872 - 21981431 on Build GRCh38 | Gain |
|
| esv2751992 | Chr.3:21745401 - 22170865 on Build GRCh38 | Gain |
|
| esv3893671 | Chr.3:21918975 - 22242630 on Build GRCh38 | Loss |
|
| nsv998319 | Chr.3:21957988 - 21991872 on Build GRCh38 | Gain |
|
| esv3568639 | Chr.3:21671375 - 22118657 on Build GRCh38 | Loss |
|
| nsv589932 | Chr.3:21943925 - 22006442 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map