Assay Details
Target Gene Details
Entrez Gene ID: | 26228 |
Gene Name: | signal transducing adaptor family member 1 |
Gene Aliases: |
BRDG1, STAP-1 |
Location: |
Chr.4:67558697-67607341 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STAP1 | NM_001317769.1 | NP_001304698.1 | ||
| NM_012108.3 | NP_036240.1 | |||
| AB023483.1 | BAA85311.1 | |||
| AK313676.1 | ||||
| BC014958.1 | AAH14958.1 |
More Information
Set Membership: |
|

Genomic Map