Assay Details
Target Gene Details
Entrez Gene ID: | 9848 |
Gene Name: | microfibrillar associated protein 3 like |
Gene Aliases: |
NYD-sp9 |
Location: |
Chr.4:169986597-170033028 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MFAP3L | NM_001301647.1 | NP_001288576.1 | ||
| NM_021647.7 | NP_067679.6 | |||
| XM_005263366.3 | XP_005263423.1 | |||
| XM_017008864.1 | 2 | 99 | XP_016864353.1 | |
| XM_017008865.1 | 2 | 3215 | XP_016864354.1 | |
| XM_017008866.1 | 2 | 289 | XP_016864355.1 | |
| XM_017008867.1 | 2 | 95 | XP_016864356.1 | |
| XM_017008868.1 | 2 | 289 | XP_016864357.1 | |
| XM_017008869.1 | XP_016864358.1 | |||
| XM_017008870.1 | XP_016864359.1 | |||
| XM_017008871.1 | XP_016864360.1 | |||
| AB014526.1 | ||||
| AK290041.1 | ||||
| BC001279.1 | 2 | 428 | AAH01279.1 | |
| DA324165.1 | ||||
| DA412844.1 |
More Information
Set Membership: |
|

Genomic Map