Assay Details
Target Gene Details
Entrez Gene ID: | 4547 |
Gene Name: | microsomal triglyceride transfer protein |
Gene Aliases: |
ABL, MTP |
Location: |
Chr.4:99564078-99623997 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTTP | NM_000253.3 | NP_000244.2 | ||
| NM_001300785.1 | NP_001287714.1 | |||
| AK225328.1 | ||||
| AK290793.1 | ||||
| AK302602.1 | ||||
| BC125110.1 | ||||
| BC125111.1 | ||||
| X59657.1 | CAA42200.1 | |||
| X75500.1 | CAA53217.1 | |||
| X91148.1 | CAA62585.1 |
More Information
Set Membership: |
|

Genomic Map