Assay Details
Target Gene Details
Entrez Gene ID: | 347732 |
Gene Name: | cation channel sperm associated 3 |
Gene Aliases: |
CACRC |
Location: |
Chr.5:134967906-135011707 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CATSPER3 | NM_178019.2 | NP_821138.1 | ||
| AF432876.1 | AAO85416.1 | |||
| AK292427.1 | ||||
| BC101692.1 | AAI01693.1 | |||
| BC110384.1 | ||||
| BI827754.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519879 | Chr.5:135007030 - 135012266 on Build GRCh38 | Loss |
|
| nsv5013 | Chr.5:134976229 - 135008381 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map