Assay Details
Target Gene Details
Entrez Gene ID: | 285600 |
Gene Name: | KIAA0825 |
Gene Aliases: |
C5orf36 |
Location: |
Chr.5:94150851-94618604 on Build GRCh38 |
Assay Gene Location: | Within Intron 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIAA0825 | NM_001145678.1 | NP_001139150.1 | ||
| XM_006714588.2 | XP_006714651.1 | |||
| AK130941.1 | BAC85469.1 |

Genomic Map