Assay Details
Target Gene Details
Entrez Gene ID: | 2697 |
Gene Name: | gap junction protein alpha 1 |
Gene Aliases: |
AVSD3, CMDR, CX43, EKVP, GJAL, HLHS1, HSS, ODDD, PPKCA |
Location: |
Chr.6:121435577-121449744 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GJA1 | NM_000165.4 | NP_000156.1 | ||
AK223289.1 | BAD97009.1 | |||
AK294670.1 | ||||
AK295460.1 | ||||
AK297402.1 | ||||
AK297761.1 | ||||
AK297764.1 | ||||
AK309377.1 | ||||
AK309490.1 | ||||
AK312324.1 | ||||
BC026329.1 | AAH26329.1 | |||
BP226987.1 | ||||
M65188.1 | AAA52131.1 | |||
X52947.1 | CAA37122.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3890934 | Chr.6:121281000 - 121612572 on Build GRCh38 | Gain |
![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
