Assay Details
Target Gene Details
Entrez Gene ID: | 3066 |
Gene Name: | histone deacetylase 2 |
Gene Aliases: |
HD2, RPD3, YAF1 |
Location: |
Chr.6:113936156-113971195 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HDAC2 | NM_001527.3 | 14 | 5441 | NP_001518.3 |
| NR_033441.1 | 15 | 5469 | ||
| NR_073443.1 | 14 | 5347 |
More Information
Set Membership: |
|

Genomic Map