Assay Details
Target Gene Details
Entrez Gene ID: | 221421 |
Gene Name: | radial spoke head 9 homolog |
Gene Aliases: |
C6orf206, CILD12, MRPS18AL1 |
Location: |
Chr.6:43645030-43671011 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RSPH9 | NM_001193341.1 | NP_001180270.1 | ||
| NM_152732.4 | NP_689945.2 | |||
| XM_005248901.3 | XP_005248958.1 | |||
| AK055407.1 | BAB70918.1 | |||
| AK291399.1 | ||||
| BC029519.1 | AAH29519.1 |
More Information
Set Membership: |
|

Genomic Map