Assay Details
Target Gene Details
Entrez Gene ID: | 64928 |
Gene Name: | mitochondrial ribosomal protein L14 |
Gene Aliases: |
L14mt, L32mt, MRP-L14, MRP-L32, MRPL32, RMPL32, RPML32 |
Location: |
Chr.6:44113454-44127491 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL14 | NM_001318767.1 | NP_001305696.1 | ||
| NM_001318768.1 | NP_001305697.1 | |||
| NM_001318769.1 | NP_001305698.1 | |||
| NM_001318770.1 | NP_001305699.1 | |||
| NM_001318771.1 | NP_001305700.1 | |||
| NM_032111.3 | NP_115487.2 | |||
| AK312086.1 | ||||
| BG545603.1 | ||||
| BU597168.1 | ||||
| BU753526.1 | ||||
| CF541260.1 | ||||
| CK823336.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv462935 | Chr.6:44078406 - 44149870 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map