Assay Details
Target Gene Details
Entrez Gene ID: | 6732 |
Gene Name: | SRSF protein kinase 1 |
Gene Aliases: |
SFRSK1 |
Location: |
Chr.6:35833034-35921180 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRPK1 | NM_003137.4 | NP_003128.3 | ||
| NR_034069.1 | ||||
| AJ318054.1 | CAC39299.1 | |||
| AK292277.1 | ||||
| AK299591.1 | ||||
| AK312712.1 | ||||
| BC038292.1 | AAH38292.1 | |||
| BX648663.1 | ||||
| BX648697.1 |
More Information
Set Membership: |
|

Genomic Map