Assay Details
Target Gene Details
Entrez Gene ID: | 3354 |
Gene Name: | 5-hydroxytryptamine receptor 1E |
Gene Aliases: |
5-HT1E |
Location: |
Chr.6:86936919-87020068 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HTR1E | NM_000865.2 | NP_000856.1 | ||
| XM_011535789.2 | XP_011534091.1 | |||
| AL521170.3 | ||||
| BC069751.1 | AAH69751.1 | |||
| M91467.1 | AAA58353.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv604106 | Chr.6:85995537 - 87138584 on Build GRCh38 | Gain |
|
| esv2763585 | Chr.6:85938269 - 87211344 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74559531] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map