Assay Details
Target Gene Details
Entrez Gene ID: | 8875 |
Gene Name: | vanin 2 |
Gene Aliases: |
FOAP-4, GPI-80 |
Location: |
Chr.6:132743503-132763489 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VNN2 | NM_078488.2 | NP_511043.1 | ||
| XM_006715593.3 | XP_006715656.1 | |||
| XM_017011407.1 | XP_016866896.1 | |||
| XM_017011408.1 | XP_016866897.1 | |||
| XM_017011409.1 | XP_016866898.1 | |||
| AK292125.1 | ||||
| DC412933.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3610846 | Chr.6:132756695 - 132762169 on Build GRCh38 | Gain |
|
| dgv1219e214 | Chr.6:132754654 - 132762349 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map