Assay Details
Target Gene Details
Entrez Gene ID: | 9774 |
Gene Name: | BCL2 associated transcription factor 1 |
Gene Aliases: |
BTF, bK211L9.1 |
Location: |
Chr.6:136256863-136290645 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCLAF1 | NM_001077440.1 | NP_001070908.1 | ||
| NM_001077441.1 | NP_001070909.1 | |||
| NM_001301038.1 | NP_001287967.1 | |||
| NM_014739.2 | NP_055554.1 | |||
| XM_005267237.3 | XP_005267294.1 | |||
| XM_017011529.1 | XP_016867018.1 | |||
| XM_017011530.1 | XP_016867019.1 | |||
| XM_017011531.1 | XP_016867020.1 | |||
| XM_017011532.1 | XP_016867021.1 | |||
| AF249273.1 | AAF64304.1 | |||
| BC063846.1 | AAH63846.1 | |||
| BC132780.1 | ||||
| BC144281.1 | ||||
| BX648878.1 | ||||
| BX649160.1 | ||||
| D79986.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2732749 | Chr.6:136269546 - 136273069 on Build GRCh38 | Deletion |
|
| nsv1124119 | Chr.6:136269611 - 136271993 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map