Assay Details
Target Gene Details
Entrez Gene ID: | 54887 |
Gene Name: | UHRF1 binding protein 1 |
Gene Aliases: |
C6orf107, ICBP90, dJ349A12.1 |
Location: |
Chr.6:34792017-34877514 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UHRF1BP1 | NM_017754.3 | NP_060224.3 | ||
| AB126777.1 | BAD32740.1 | |||
| BC137120.1 | ||||
| BC137127.1 | ||||
| BX640641.1 | CAE45788.1 | |||
| DB636320.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv602906 | Chr.6:34760476 - 34918659 on Build GRCh38 | Gain |
|
| dgv5955n100 | Chr.6:34755946 - 34919539 on Build GRCh38 | Gain |
|
| esv2763967 | Chr.6:34598478 - 34957686 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map