Assay Details
Target Gene Details
Entrez Gene ID: | 56897 |
Gene Name: | Werner helicase interacting protein 1 |
Gene Aliases: |
WHIP, bA420G6.2 |
Location: |
Chr.6:2765341-2785745 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WRNIP1 | NM_020135.2 | NP_064520.2 | ||
| NM_130395.2 | NP_569079.1 | |||
| AB056152.1 | BAB60709.1 | |||
| AB209723.1 | BAD92960.1 | |||
| AF218313.1 | AAF80563.1 | |||
| AK026179.1 | BAB15383.1 | |||
| AK094823.1 | ||||
| AK223593.1 | BAD97313.1 | |||
| AK315047.1 | ||||
| AK315471.1 | ||||
| BC018923.2 | AAH18923.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3607900 | Chr.6:2093592 - 2775672 on Build GRCh38 | Gain |
|
| esv3607912 | Chr.6:2623291 - 2776818 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map