Assay Details
Target Gene Details
Entrez Gene ID: | 116113 |
Gene Name: | forkhead box P4 |
Gene Aliases: |
hFKHLA |
Location: |
Chr.6:41546426-41602384 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FOXP4 | NM_001012426.1 | NP_001012426.1 | ||
| NM_001012427.1 | NP_001012427.1 | |||
| NM_138457.2 | NP_612466.1 | |||
| XM_006714991.3 | XP_006715054.1 | |||
| XM_011514289.2 | XP_011512591.1 | |||
| XM_011514290.2 | XP_011512592.1 | |||
| XM_011514291.2 | XP_011512593.1 | |||
| XM_011514292.2 | XP_011512594.1 | |||
| XM_017010234.1 | XP_016865723.1 | |||
| BC040962.1 | AAH40962.1 | |||
| BC052803.1 | AAH52803.1 |
Target Gene Details
Entrez Gene ID: | 101060264 |
Gene Name: | FOXP4 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.6:41523895-41548621 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FOXP4-AS1 | NR_126415.1 | |||
| NR_126416.1 | ||||
| BF194903.1 | ||||
| BQ128057.1 | ||||
| BQ581966.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1073551 | Chr.6:41546161 - 41548062 on Build GRCh38 | Deletion |
|
| nsv1144141 | Chr.6:41546162 - 41547362 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map