Assay Details
Target Gene Details
Entrez Gene ID: | 60682 |
Gene Name: | small ArfGAP 1 |
Gene Aliases: |
SMAP-1 |
Location: |
Chr.6:70667771-70862015 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMAP1 | NM_001044305.2 | NP_001037770.1 | ||
| NM_001281439.1 | NP_001268368.1 | |||
| NM_001281440.1 | NP_001268369.1 | |||
| NM_021940.4 | NP_068759.2 | |||
| XM_005248760.4 | XP_005248817.1 | |||
| XM_011536045.2 | XP_011534347.1 | |||
| XM_011536046.2 | XP_011534348.1 | |||
| XM_011536047.2 | XP_011534349.1 | |||
| XM_011536048.2 | XP_011534350.1 | |||
| XM_017011170.1 | XP_016866659.1 | |||
| AF442495.1 | AAP97320.1 | |||
| AK023221.1 | BAB14473.1 | |||
| AK124239.1 | ||||
| AK222711.1 | BAD96431.1 | |||
| AK290448.1 | ||||
| AK303387.1 | ||||
| AK307807.1 | ||||
| AK315579.1 | ||||
| AY055003.1 | AAL14714.1 | |||
| AY055004.1 | AAL14715.1 | |||
| BC008672.2 | AAH08672.1 | |||
| BC028074.1 | AAH28074.1 | |||
| BC036123.1 | AAH36123.1 | |||
| DA527013.1 | ||||
| DB480952.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524328 | Chr.6:70252130 - 70849102 on Build GRCh38 | Gain |
|
| nsv1027049 | Chr.6:70251907 - 70851785 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map