Assay Details
Target Gene Details
Entrez Gene ID: | 25862 |
Gene Name: | ubiquitin specific peptidase 49 |
Gene Aliases: |
- |
Location: |
Chr.6:41789896-41895361 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP49 | NM_001286554.1 | 6 | 8069 | NP_001273483.1 |
| AK023682.1 | 1 | 1776 | ||
| AL110255.1 | 1 | 1417 |
More Information
Set Membership: |
|

Genomic Map