Assay Details
Target Gene Details
Entrez Gene ID: | 8763 |
Gene Name: | CD164 molecule |
Gene Aliases: |
DFNA66, MGC-24, MUC-24, endolyn |
Location: |
Chr.6:109366514-109382559 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD164 | NM_001142401.1 | NP_001135873.1 | ||
| NM_001142402.1 | NP_001135874.1 | |||
| NM_001142403.1 | NP_001135875.1 | |||
| NM_001142404.1 | NP_001135876.1 | |||
| NM_006016.4 | NP_006007.2 | |||
| AF106518.1 | AAC82473.1 | |||
| AF263279.1 | AAF85965.1 | |||
| AF299341.1 | AAG53906.1 | |||
| AF299342.1 | AAG53907.1 | |||
| AF299343.1 | AAG53908.1 | |||
| AK298684.1 | ||||
| AK301692.1 | ||||
| AK303525.1 | ||||
| AK307193.1 | ||||
| AK308664.1 | ||||
| AK310854.1 | ||||
| AK312357.1 | ||||
| AK315908.1 | ||||
| BC011522.2 | AAH11522.1 | |||
| D14043.1 | BAA03130.1 | |||
| FJ200494.1 |
More Information
Set Membership: |
|

Genomic Map