Assay Details
Target Gene Details
Entrez Gene ID: | 100130357 |
Gene Name: | uncharacterized LOC100130357 |
Gene Aliases: |
- |
Location: |
Chr.6:13273094-13295586 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100130357 | NR_134859.1 | |||
| BX089972.1 |
Target Gene Details
Entrez Gene ID: | 221692 |
Gene Name: | phosphatase and actin regulator 1 |
Gene Aliases: |
RPEL, RPEL1, dJ257A7.2 |
Location: |
Chr.6:12716635-13290484 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
More Information
Set Membership: |
|

Genomic Map