Assay Details
Target Gene Details
Entrez Gene ID: | 135228 |
Gene Name: | CD109 molecule |
Gene Aliases: |
CPAMD7, p180, r150 |
Location: |
Chr.6:73679192-73828317 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD109 | NM_001159587.2 | NP_001153059.1 | ||
| NM_001159588.2 | NP_001153060.1 | |||
| NM_133493.4 | NP_598000.2 | |||
| XM_005248659.3 | XP_005248716.1 | |||
| XM_011535473.1 | XP_011533775.1 | |||
| AF410459.1 | AAL84159.1 | |||
| AK095888.1 | BAC04642.1 | |||
| AK313636.1 | ||||
| AY149920.1 | AAN78483.1 | |||
| AY788891.1 | ||||
| BX641095.1 | CAE46045.1 | |||
| EF553520.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv463154 | Chr.6:73702325 - 74066188 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map