Assay Details
Target Gene Details
Entrez Gene ID: | 23070 |
Gene Name: | cap methyltransferase 1 |
Gene Aliases: |
FTSJD2, KIAA0082, MTr1, hMTr1 |
Location: |
Chr.6:37433131-37482844 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CMTR1 | NM_015050.2 | NP_055865.1 | ||
| XM_005248955.3 | XP_005249012.1 | |||
| AK292564.1 | ||||
| BC031890.1 | AAH31890.1 | |||
| D43949.2 | ||||
| DA508836.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1015689 | Chr.6:36550916 - 39052434 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map