Assay Details
Target Gene Details
Entrez Gene ID: | 222553 |
Gene Name: | solute carrier family 35 member F1 |
Gene Aliases: |
C6orf169, dJ230I3.1 |
Location: |
Chr.6:117907226-118317676 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC35F1 | NM_001029858.3 | NP_001025029.2 | ||
| XM_005266865.4 | XP_005266922.1 | |||
| BC028615.1 | AAH28615.1 | |||
| BC036493.1 | AAH36493.1 | |||
| BC114925.1 | AAI14926.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522455 | Chr.6:117907364 - 117919522 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map