Assay Details
Target Gene Details
Entrez Gene ID: | 4201 |
Gene Name: | male-enhanced antigen 1 |
Gene Aliases: |
HYS, MEA |
Location: |
Chr.6:42980344-43016886 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MEA1 | XM_017010868.1 | XP_016866357.1 | ||
| AK309759.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3576145 | Chr.6:42728097 - 43012090 on Build GRCh38 | Gain |
|
| nsv830651 | Chr.6:42845563 - 43000871 on Build GRCh38 | Loss |
|
| nsv462927 | Chr.6:42948182 - 43110718 on Build GRCh38 | Loss |
|
| nsv1017398 | Chr.6:42659705 - 43400998 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map