Assay Details
Target Gene Details
Entrez Gene ID: | 101927365 |
Gene Name: | uncharacterized LOC101927365 |
Gene Aliases: |
- |
Location: |
Chr.6:99424871-99431315 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927365 | NR_125843.1 | |||
| AK025621.1 | 1 | 592 | ||
| DB459887.1 |
Target Gene Details
Entrez Gene ID: | 100130890 |
Gene Name: | thiosulfate sulfurtransferase (rhodanese)-like domain containing 3 |
Gene Aliases: |
- |
Location: |
Chr.6:99424854-99579288 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSTD3 | XM_017010141.1 | 1 | 699 | XP_016865630.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv823789 | Chr.6:99424816 - 99426378 on Build GRCh38 | Loss |
|
| nsv1029536 | Chr.6:98422726 - 99818247 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map